missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-phenylbutanoic Acid, TRC

Product Code. 30347940
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30347940 100 mg 100mg
30337694 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30347940 Supplier Toronto Research Chemicals Supplier No. TRCA630595100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Amino-3-phenylbutanoic Acid
CAS 1078-21-3
Purity Grade Notes HPLC
Molecular Formula C10H13NO2
Synonym -(Aminomethyl)benzenepropanoic Acid, (±)-Fenibut, (±)-beta-Phenyl-GABA, 3-Phenyl-4-aminobutanoic Acid, 4-Amino-3-phenylbutyric Acid, Anvifen, DL-4-Amino-3-phenylbutanoic Acid, DL-beta-Phenyl-gamma-aminobutyric Acid, Fenibut, Fenigam, Fenigama, P-GABA, PhGABA, Phenibut, Phenigam, Phenybut, Phenygam, beta-Phenyl-GABA, beta-Phenyl-gamma-aminobutyric Acid, gamma-Amino-beta-phenylbutyric Acid
InChI Formula InChI=1S/C10H13NO2/c11-7-9(6-10(12)13)8-4-2-1-3-5-8/h1-5,9H,6-7,11H2,(H,12,13)
SMILES NCC(CC(=O)O)c1ccccc1
IUPAC Name 4-amino-3-phenylbutanoic acid
Molecular Weight (g/mol) 179.22
Quantity 100 mg
Formula Weight 179.0946
Percent Purity >95
Shipping Condition Room Temperature
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.