missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitropyridine, 96%

Product Code. 11375565
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11375565 5 g 5g
11365565 1 g 1g
11385565 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11375565 Supplier Thermo Scientific Alfa Aesar Supplier No. L19725.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1681-37-4 | C5H5N3O2 | 139.114 g/mol

4-Amino-3-nitropyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-nitropyridine is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1681-37-4
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.114
MDL Number MFCD00160304
InChI Key IUPPEELMBOPLDJ-UHFFFAOYSA-N
Synonym 4-amino-3-nitropyridine, 4-pyridinamine, 3-nitro, 3-nitro-4-pyridinamine, 3-nitro-4-aminopyridine, 3-nitro-pyridin-4-ylamine, 3-nitro-4-pyridylamine, pubchem1277, acmc-209dxo, 3-nitro-4-amino pyridine, 3-nitro-4-pyridinamine #
PubChem CID 548803
IUPAC Name 3-nitropyridin-4-amine
SMILES C1=CN=CC(=C1N)[N+](=O)[O-]

Specifications

Melting Point 201°C to 205°C
Quantity 5 g
Beilstein 383710
Solubility Information Insoluble in water.
Formula Weight 139.11
Percent Purity 96%
Chemical Name or Material 4-Amino-3-nitropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.