missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitrophenol, 98%

Product Code. 10071721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071721 25 g Glass bottle 25g
10215481 5 g Glass bottle 5g
10552544 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10071721 Supplier Thermo Scientific Acros Supplier No. 335220250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 610-81-1 | C6H6N2O3 | 154.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-81-1
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00066310
InChI Key IQXUIDYRTHQTET-UHFFFAOYSA-N
Synonym 4-hydroxy-2-nitroaniline, 3-nitro-4-aminophenol, phenol, 4-amino-3-nitro, 4-amino-3-nitro-phenol, unii-r5wy41q95z, 3-nitro-4-amino phenol, 4-aminophenol-3-nitrophenol, pubchem1481, 4-amino-3-nitropbenol
PubChem CID 3758882
IUPAC Name 4-amino-3-nitrophenol
SMILES C1=CC(=C(C=C1O)[N+](=O)[O-])N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >150.0°C
Color Bordeaux to Red
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 13, III, 1203
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, isopropanol,, acetone, dimethyl sulfoxide, chloroform,, methylene chloride, xylene, benzene, diethyl ether
Formula Weight 154.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-nitrophenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Amino-3-nitrophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.