missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-methylbenzoic acid, 98%

Product Code. p-7027655
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11400428

Brand: Thermo Scientific Alfa Aesar B24579.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2486-70-6 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2486-70-6
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00007736
InChI Key NHFKECPTBZZFBC-UHFFFAOYSA-N
Synonym 3-methyl-4-aminobenzoic acid, 4-amino-m-toluic acid, benzoic acid, 4-amino-3-methyl, 4-amino-3-methyl benzoic acid, 4-amino-3-methyl benzoate, 4-amino-3-methylbenzoicacid, unii-201cxt4h5v, 4-amino-3-methyl-benzoic acid, benzoic acid, 3-methyl-4-amino, 4-amino-3-methylbenzenecarboxylic acid
PubChem CID 75598
IUPAC Name 4-amino-3-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)C(=O)O)N

Specifications

Melting Point 169°C to 171°C
Quantity 1 g
Beilstein 2802615
Formula Weight 151.16
Percent Purity 98%
Chemical Name or Material 4-Amino-3-methylbenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.