missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-methoxybenzoic acid, 98%

Product Code. 10390144 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10390144 5 g Glass bottle 5g
10792284 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10390144 Supplier Thermo Scientific Acros Supplier No. 391460050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2486-69-3 | C8H9NO3 | 167.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2486-69-3
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD00016539
InChI Key JNFGLYJROFAOQP-UHFFFAOYSA-N
Synonym 4-amino-3-methoxy-benzoic acid, 3-methoxy-4-aminobenzoic acid, 4-amino-m-anisic acid, benzoic acid, 4-amino-3-methoxy, o-anisidine-4-carboxylic acid, 4-amino-3-methoxy benzoic acid, timtec-bb sbb008622, 4-amino-3-methoxybenzoic, 4-amino-3-methoxybenzoic acid, 4-amino-3-methoxybenzoicacid
PubChem CID 288057
IUPAC Name 4-amino-3-methoxybenzoic acid
SMILES COC1=C(C=CC(=C1)C(=O)O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 186°C to 188°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3OC6H3(NH2)COOH
Quantity 5 g
Formula Weight 167.16
Percent Purity 98%
Physical Form Powder, Crystals and/or Chunks or Fused Solid
Chemical Name or Material 4-Amino-3-methoxybenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Amino-3-methoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.