missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-iodobenzoic acid, 98%, Thermo Scientific™

Product Code. 15454967
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15454967

Brand: Thermo Scientific Alfa Aesar H32263.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Resin-bound 3-[2-(acetylamino)ethyl]-2- iodo-1H-indole-5-carboxamide was generated starting from Polystyrene- Rink resin and 4-amino-3-iodobenzoic acid.

Solubility
Soluble in water(50 g/L) (25°C).

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light. Store in the dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2122-63-6
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.03
MDL Number MFCD01660550
InChI Key FPLDDZRJTOXFCB-UHFFFAOYSA-N
Synonym benzoic acid, 4-amino-3-iodo, 4-amino-3-iodobenzoicacid, 4-amino-3-iodo-benzoic acid, pubchem14046, rarechem al bo 1498, 4-carboxy-2-iodoaniline, 4-amino-3-iodo benzoic, wln: zr bi dvq, 4-amino-3-iodo-benzoicaci, 3-iodo-4-aminobenzoic acid
PubChem CID 16461
IUPAC Name 4-amino-3-iodobenzoic acid
SMILES NC1=CC=C(C=C1I)C(O)=O

Specifications

Quantity 5 g
Sensitivity Light sensitive
Solubility Information Soluble in water(50g/L) (25°C).
Formula Weight 263.03
Percent Purity 98%
Chemical Name or Material 4-Amino-3-iodobenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.