missing translation for 'onlineSavingsMsg'
Läs mer

4-Amino-3-hydroxybenzoic acid, 98%

Produktkod. 11419532
Change view
Klicka för att se tillgängliga alternativ
Quantity:
5 g
25 g
100 g
Förpackningsstorlek:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
11419532 100 g 100g
11499522 5 g 5g
11409532 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11419532 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer A10207.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 2374-03-0 | C7H7NO3 | 153.137 g/mol

4-Amino-3-hydroxybenzoic acid is used in the preparation of various pharmaceutical compounds such as sphingosine kinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-hydroxybenzoic acid is used in the preparation of various pharmaceutical compounds such as sphingosine kinase inhibitors.

Solubility
Soluble in chloroform, and aqeous ethanol, water (Partly miscible)

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 2374-03-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00017094
InChI Key NFPYJDZQOKCYIE-UHFFFAOYSA-N
Synonym 3-hydroxy-4-aminobenzoic acid, 4-amino-3-hydroxy benzoic acid, benzoic acid, 4-amino-3-hydroxy, chembl1462, 2hdr, pubchem14418, 2-amino-5-carboxyphenol, intermediates-zcf02635, acmc-209g5k, 4-carboxy-2-hydroxyaniline
PubChem CID 137566
IUPAC Name 4-amino-3-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)O)N

Specifikationer

Melting Point ∼215°C (decomposition)
Quantity 100 g
Beilstein 509859
Solubility Information Soluble in chloroform,and aqeous ethanol,water (Partly miscible)
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material 4-Amino-3-hydroxybenzoic acid

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.