missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-ethylbenzonitrile, 96%

Product Code. 11362966
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362966 1 g 1g
11372966 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362966 Supplier Thermo Scientific Alfa Aesar Supplier No. L10732.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 170230-87-2 | C9H10N2 | 146.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170230-87-2
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.193
MDL Number MFCD00041431
InChI Key YOHLABDNVQLZIA-UHFFFAOYSA-N
Synonym 4-amino-3-ethyl-benzonitrile, 4-cyano-2-ethylaniline, 4-amino-3-ethyl benzonitrile, buttpark 13\03-08, benzonitrile, 4-amino-3-ethyl-9ci, 4-amino-3-ethylbenzenecarbonitrile, pubchem16098, pubchem21039, acmc-209xcd, intermediates-zcf02269
PubChem CID 2735330
IUPAC Name 4-amino-3-ethylbenzonitrile
SMILES CCC1=C(C=CC(=C1)C#N)N

Specifications

Melting Point 60°C to 66°C
Quantity 1 g
UN Number UN3439
Beilstein 7526452
Formula Weight 146.19
Percent Purity 96%
Chemical Name or Material 4-Amino-3-ethylbenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.