missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-chlorobenzotrifluoride, 97%, ACROS Organics™

Product Code. 10409671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10409671 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10409671 Supplier Thermo Scientific Acros Supplier No. 326440050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 39885-50-2
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H5ClF3N
Refractive Index 1.5010 to 1.5040 (20°C, 589nm)
MDL Number MFCD00042563
Quantity 5 g
Synonym 2-chloro-4-trifluoromethyl aniline, 4-amino-3-chlorobenzotrifluoride, 4-amino-5-chloro-benzotrifluoride, benzenamine, 2-chloro-4-trifluoromethyl, 3-chloro-4-aminobenzotrifluoride, 2-chloro-4-trifluoromethyl-phenylamine, 2-chloro-4-trifluoromethyl benzenamine, 2-chloro-4-trifluoromethyl phenylamine, 4-amino-3-chloro-benzotrifluoride, pubchem2474
InChI Key MBBUTABXEITVNY-UHFFFAOYSA-N
SMILES NC1=CC=C(C=C1Cl)C(F)(F)F
IUPAC Name 2-chloro-4-(trifluoromethyl)aniline
Molecular Weight (g/mol) 195.57
PubChem CID 162001
Formula Weight 195.57
Percent Purity ≥96.0%
Physical Form Liquid After Melting
Chemical Name or Material 4-Amino-3-chlorobenzotrifluoride, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.