missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-bromobenzonitrile, 97%, Thermo Scientific Chemicals

Product Code. 15401758
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50397-74-5
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD01861386
InChI Key POESQIHWIIWNJL-UHFFFAOYSA-N
Synonym 3-bromo-4-aminobenzonitrile, 2-bromo-4-cyanoaniline, benzonitrile, 4-amino-3-bromo, 4-amino-3-bromo-benzonitrile, 4-amino-3-brombenzonitrile, buttpark 35\03-64, 4-amino-3-bromobenzenecarbonitrile, pubchem4609, acmc-209klz, 4-amino-3-bromobenzonitile
PubChem CID 1515279
IUPAC Name 4-amino-3-bromobenzonitrile
SMILES C1=CC(=C(C=C1C#N)Br)N

Specifications

Melting Point 106°C to 110°C
Quantity 250 mg
UN Number UN3439
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 4-Amino-3-bromobenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'