missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3,5-xylenol, TRC

Product Code. 30329157
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329157 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329157 Supplier Toronto Research Chemicals Supplier No. TRCA63243510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Amino-3,5-xylenol
CAS 3096-70-6
Molecular Formula C8H11NO
Quantity 10 g
Synonym 4-Amino-3,5-dimethyl-phenol, 3,5-Dimethyl-4-aminophenol, 4-Hydroxy-2,6-dimethylaniline, 4-Hydroxy-2,6-xylidine, NSC 38030
InChI Formula InChI=1S/C8H11NO/c1-5-3-7(10)4-6(2)8(5)9/h3-4,10H,9H2,1-2H3
SMILES CC1=C(C(=CC(=C1)O)C)N
IUPAC Name 4-amino-3,5-dimethylphenol
Molecular Weight (g/mol) 137.18
Formula Weight 137.0841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.