missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-picoline, 98+%, Thermo Scientific™

Product Code. 10269333 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10269333 5 g Glass bottle 5g
10320344 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10269333 Supplier Thermo Scientific Acros Supplier No. 393650050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 18437-58-6
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00186509
InChI Key GNCLPNMQEGMNTG-UHFFFAOYSA-N
Synonym 4-amino-2-methylpyridine, 4-amino-2-picoline, 2-methyl-4-aminopyridine, 2-methyl-4-pyridinamine, 4-pyridinamine, 2-methyl, 2-methyl-pyridin-4-ylamine, 4 1h-pyridinimine, 2-methyl, 4-amino-picoline, 4-amino-6-picoline, pubchem1118
PubChem CID 728670
IUPAC Name 2-methylpyridin-4-amine
SMILES CC1=NC=CC(=C1)N

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 92°C to 95°C
Color Tan
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 108.14
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 4-Amino-2-picoline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Amino-2-picoline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.