missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Amino-2-nitrophenol, 97%

Product Code. 11382789
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382789 5 g 5g
11392789 25 g 25g
11302799 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11382789 Supplier Alfa Aesar™ Supplier No. H27694.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
4-Amino-2-nitrophenol was used in determination of aminonitrophenols in hair dyes by differential pulse voltammetry and HPLC with electrochemical detection. It is commonly used as semipermanent (nonoxidative) hair colorant and toner in permanent (oxidative) hair dye products.

Solubility
Slightly soluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-34-6
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00007876
InChI Key WHODQVWERNSQEO-UHFFFAOYSA-N
Synonym phenol, 4-amino-2-nitro, fourrine 57, 4-hydroxy-3-nitroaniline, 2-nitro-4-aminophenol, fourrine brown pr, oxidation base 25, fourrine brown propyl, o-nitro-p-aminophenol, p-amino-o-nitrophenol, c.i. oxidation base 25
PubChem CID 3417419
ChEBI CHEBI:82385
IUPAC Name 4-amino-2-nitrophenol
SMILES C1=CC(=C(C=C1N)[N+](=O)[O-])O

Specifications

Melting Point 127°C to 130°C
Quantity 5 g
Beilstein 1368435
Formula Weight 154.13
Percent Purity 97%
Chemical Name or Material 4-Amino-2-nitrophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.