missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-(methylthio)pyrimidine-5-carbonitrile, 97%

Product Code. 11488807
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488807 1 g 1g
11498807 5 g 5g
11408817 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488807 Supplier Thermo Scientific Alfa Aesar Supplier No. A19949.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 770-30-9 | C6H6N4S | 166.20 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 770-30-9
Molecular Formula C6H6N4S
Molecular Weight (g/mol) 166.20
MDL Number MFCD00023237
InChI Key VYDHBDPZNLVELI-UHFFFAOYSA-N
Synonym 4-amino-2-methylthio pyrimidine-5-carbonitrile, 4-amino-2-methylsulfanyl pyrimidine-5-carbonitrile, 4-amino-2-methylthio-5-pyrimidinecarbonitrile, 4-amino-2-methylsulfanyl-pyrimidine-5-carbonitrile, 4-amino-2-methylthiopyrimidine-5-carbonitrile, maybridge1_004789, acmc-20a4j5, 4-amino-5-cyano-2-methylthiopyrimidine, 4-amino-5-cyano-2-methylthio pyrimidine
PubChem CID 295757
IUPAC Name 4-amino-2-methylsulfanylpyrimidine-5-carbonitrile
SMILES CSC1=NC=C(C#N)C(N)=N1

Specifications

Melting Point 240°C to 243°C
Quantity 1 g
UN Number UN3439
Solubility Information Soluble in water.
Formula Weight 166.21
Percent Purity 97%
Chemical Name or Material 4-Amino-2-(methylthio)pyrimidine-5-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.