missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-methylpyridine, 98%

Product Code. 15451158
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451158 250 mg 250mg
15431158 1 g 1g
15441158 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15451158 Supplier Thermo Scientific Alfa Aesar Supplier No. H55036.MD
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18437-58-6 | C6H8N2 | 108.144 g/mol

4-Amino-2-methylpyridine is used to prepare 2-methyl-pyridin-4-ol; nitrate at 20°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-2-methylpyridine is used to prepare 2-methyl-pyridin-4-ol; nitrate at 20°C.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18437-58-6
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00186509
InChI Key GNCLPNMQEGMNTG-UHFFFAOYSA-N
Synonym 4-amino-2-methylpyridine, 4-amino-2-picoline, 2-methyl-4-aminopyridine, 2-methyl-4-pyridinamine, 4-pyridinamine, 2-methyl, 2-methyl-pyridin-4-ylamine, 4 1h-pyridinimine, 2-methyl, 4-amino-picoline, 4-amino-6-picoline, pubchem1118
PubChem CID 728670
IUPAC Name 2-methylpyridin-4-amine
SMILES CC1=NC=CC(=C1)N

Specifications

Melting Point 93°C to 97°C
Quantity 250 mg
Solubility Information Soluble in methanol.
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material 4-Amino-2-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.