missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-hydroxypyridine, 97%

Product Code. 11996341 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11996341 1 g Glass bottle 1g
11906351 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11996341 Supplier Thermo Scientific Acros Supplier No. 432080010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38767-72-5 | C5H6N2O | 110.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38767-72-5
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
InChI Key SBQVQYPJWLJRQT-UHFFFAOYSA-N
PubChem CID 573530
IUPAC Name 4-amino-1H-pyridin-2-one
SMILES C1=CNC(=O)C=C1N

Specifications

Melting Point 206.5°C to 208.5°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Synonym 4-aminopyridin-2-ol, 4-amino-2-hydroxypyridine, 4-amino-pyridin-2-ol, 4-aminopyridin-2 1h-one, 4-amino-2-pyridone, 3-deazacytosine, 2 1h-pyridinone, 4-amino, 2 1h-pyridinone,4-amino, 2-pyridinol, 4-amino, 4-amino-1,2-dihydropyridin-2-one
Formula Weight 110.12
Percent Purity 97%
Chemical Name or Material 4-Amino-2-hydroxypyridine
compliance-icons
Product Identifier
  • 4-Amino-2-hydroxypyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.