missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2-chloropyridine, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 4-Amino-2-chloropyridine |
| CAS | 14432-12-3 |
| Molecular Formula | C5 H5 Cl N2 |
| Quantity | 25 g |
| Synonym | 4-Amino-2-chloropyridine, 4-Pyridinamine, 2-chloro-, Pyridine, 4-amino-2-chloro- (6CI,7CI,8CI), 2-Chloro-4-pyridinamine, (2-Chloropyridin-4-yl)amine, 2-Chloro-4-aminopyridine, 2-Chloropyridin-4-amine, 4-Amino-2-chloropyridine, (2-Chloropyridin-4-yl)amine, 2-Chloro-4-aminopyridine, 2-Chloropyridin-4-amine, 2-Chloro-4-pyridinamine |
| InChI Formula | InChI=1S/C5H5ClN2/c6-5-3-4(7)1-2-8-5/h1-3H,(H2,7,8) |
| SMILES | Nc1ccnc(Cl)c1 |
| IUPAC Name | 2-chloropyridin-4-amine |
| Molecular Weight (g/mol) | 128.56 |
| Formula Weight | 128.0141 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction