missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chlorobenzonitrile, 99%

Product Code. 13501440 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13501440 10 g 10g
13511620 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13501440 Supplier Thermo Scientific Acros Supplier No. 443880100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 20925-27-3 | C7H5ClN2 | 152.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20925-27-3
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD00035926
InChI Key ZFBKYGFPUCUYIF-UHFFFAOYSA-N
Synonym 3-chloro-4-cyanoaniline, 2-chloro-4-aminobenzonitrile, benzonitrile, 4-amino-2-chloro, 4-amino-2-chloro-benzonitrile, 4-amino-2-chlorobenzenecarbonitrile, pubchem4620, 4-cyano-3-chloroaniline, acmc-1cfb7, 3-chloro-4-cyanobenzenamine, ksc497i5p
PubChem CID 88728
IUPAC Name 4-amino-2-chlorobenzonitrile
SMILES NC1=CC=C(C#N)C(Cl)=C1

Specifications

Melting Point 116.0°C to 118.0°C
Quantity 10 g
Packaging Glass bottle
Assay Percent Range 98.5% min. (HPLC)
Solubility Information Solubility in water: insoluble
Formula Weight 152.58
Percent Purity 99%
Chemical Name or Material 4-Amino-2-chlorobenzonitrile
compliance-icons
Product Identifier
  • 4-Amino-2-chlorobenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.