missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chlorobenzonitrile, 97+%

Product Code. 11392807
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392807 5 g 5g
11302817 25 g 25g
11312817 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11392807 Supplier Thermo Scientific Alfa Aesar Supplier No. L15924.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20925-27-3 | C7H5ClN2 | 152.58 g/mol

4-Amino-2-chlorobenzonitrile is commonly used as an organic reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-2-chlorobenzonitrile is commonly used as an organic reagent.

Notes
Incompatible with acid chlorides, acid anhydrides and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20925-27-3
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD00035926
InChI Key ZFBKYGFPUCUYIF-UHFFFAOYSA-N
Synonym 3-chloro-4-cyanoaniline, 2-chloro-4-aminobenzonitrile, benzonitrile, 4-amino-2-chloro, 4-amino-2-chloro-benzonitrile, 4-amino-2-chlorobenzenecarbonitrile, pubchem4620, 4-cyano-3-chloroaniline, acmc-1cfb7, 3-chloro-4-cyanobenzenamine, ksc497i5p
PubChem CID 88728
IUPAC Name 4-amino-2-chlorobenzonitrile
SMILES NC1=CC=C(C#N)C(Cl)=C1

Specifications

Melting Point 116°C to 119°C
Quantity 5 g
UN Number UN3439
Beilstein 2936425
Formula Weight 152.58
Percent Purity ≥98%
Chemical Name or Material 4-Amino-2-chlorobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.