missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,6-dimethylpyrimidine, 99%

Product Code. 11469707
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469707 2.5 g 2.5g
11479707 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469707 Supplier Thermo Scientific Alfa Aesar Supplier No. B22175.88
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 461-98-3 | C6H9N3 | 123.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-98-3
Molecular Formula C6H9N3
Molecular Weight (g/mol) 123.159
MDL Number MFCD00006106
InChI Key BJJDXAFKCKSLTE-UHFFFAOYSA-N
Synonym 4-amino-2,6-dimethylpyrimidine, kyanmethin, cyanomethine, cyanmethine, kyanmethine, 2,6-dimethyl-4-pyrimidinamine, 2,4-dimethyl-6-aminopyrimidine, 6-amino-2,4-dimethylpyrimidine, 2,6-dimethyl-4-pyrimidinylamine, 4-pyrimidinamine, 2,6-dimethyl
PubChem CID 68039
IUPAC Name 2,6-dimethylpyrimidin-4-amine
SMILES CC1=CC(=NC(=N1)C)N

Specifications

Melting Point 184°C to 186°C
Quantity 2.5 g
Merck Index 14,5326
Formula Weight 123.16
Percent Purity 99%
Chemical Name or Material 4-Amino-2,6-dimethylpyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.