missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1H-pyrazolo[3,4-d]pyrimidine, 98%, Thermo Scientific™

Product Code. 11342196
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342196 250 mg 250mg
11322196 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342196 Supplier Thermo Scientific Alfa Aesar Supplier No. L03519.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Amino-1H-pyrazolo[3,4-d]pyrimidine decreases serum cholesterol markedly in rats.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2380-63-4
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00005688
InChI Key LHCPRYRLDOSKHK-UHFFFAOYSA-N
Synonym 1h-pyrazolo 3,4-d pyrimidin-4-amine, 4-aminopyrazolo 3,4-d pyrimidine, 4-amino-1h-pyrazolo 3,4-d pyrimidine, pyrazoloadenine, 1h-pyrazolo 3,4-d pyrimidin-4-ylamine, 4-aminopyrazolopyrimidine, 4 app, 4-app, adenine antimetabolite
PubChem CID 75420
IUPAC Name 1H-pyrazolo[3,4-d]pyrimidin-4-amine
SMILES NC1=C2C=NNC2=NC=N1

Specifications

Melting Point >310°C (decomposition)
Odor Odorless
Quantity 250 mg
UN Number UN2811
Beilstein 5824
Solubility Information Insoluble in water.
Formula Weight 135.13
Percent Purity 98%
Chemical Name or Material 4-Amino-1H-pyrazolo[3,4-d]pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.