missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-naphthol hydrochloride, 90%, technical

Product Code. 10246110 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10246110 5 g 5g
10564091 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10246110 Supplier Thermo Scientific Acros Supplier No. 158510050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5959-56-8 | C10H9NO·HCl | 195.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5959-56-8
Molecular Formula C10H9NO·HCl
Molecular Weight (g/mol) 195.65
MDL Number MFCD00012562
InChI Key FDBQTRARWCKEJY-UHFFFAOYSA-N
Synonym 4-amino-1-naphthol hydrochloride, 4-aminonaphthalen-1-ol hydrochloride, 1-amino-4-naphthol hydrochloride, 1-naphthalenol, 4-amino-, hydrochloride, 4-amino-1-naphtholhydrochloride, 4-aminonaphthol, chloride, pcmd-cc-pab-253, 1-naphthalenol, 4-amino-, hydrochloride 1:1, acmc-1aogt, 4-amino-1-naphthol . hcl
PubChem CID 2723858
IUPAC Name 4-aminonaphthalen-1-ol;hydrochloride
SMILES C1=CC=C2C(=C1)C(=CC=C2O)N.Cl

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 273.0°C
Color Gray to Pink
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NC10H6OH·HCl
Quantity 5 g
Beilstein 13, 667
Merck Index 10, 457
Formula Weight 195.65
Percent Purity 90%
Grade Technical
Physical Form Powder
Chemical Name or Material 4-Amino-1-naphthol hydrochloride, tech., 90%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.