missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-Boc-piperidine, 97%

Product Code. p-7030605
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15443987

Brand: Thermo Scientific Alfa Aesar H30779.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87120-72-7 | C10H21N2O2 | 201.29 g/mol

4-Amino-1-Boc-piperidine is a chemical reagent used in the preparation of pharmaceutical compounds. Used in the synthesis of bromodomain inhibitors as well as HepG2 cell cycle inhibitors used in anti-tumor therapy. Also employed in a microwave-assisted solid-phase synthesis of N-substituted piperidines via direct annulation of primary amines with resin-bound dimesylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-1-Boc-piperidine is a chemical reagent used in the preparation of pharmaceutical compounds. Used in the synthesis of bromodomain inhibitors as well as HepG2 cell cycle inhibitors used in anti-tumor therapy. Also employed in a microwave-assisted solid-phase synthesis of N-substituted piperidines via direct annulation of primary amines with resin-bound dimesylates.

Solubility
Insoluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87120-72-7
Molecular Formula C10H21N2O2
Molecular Weight (g/mol) 201.29
MDL Number MFCD01076201
InChI Key LZRDHSFPLUWYAX-UHFFFAOYSA-O
Synonym 4-amino-1-boc-piperidine, 1-boc-4-aminopiperidine, n-boc-4-aminopiperidine, n-boc-4-amino-piperidine, 4-amino-1-n-boc-piperidine, 4-amino-n-boc piperidine, 1-boc-4-amino piperidine, 1-n-boc-4-aminopiperidine, 1-t-butoxycarbonyl-4-aminopiperidine, n-1-boc-4-aminopiperidine
PubChem CID 1268291
IUPAC Name tert-butyl 4-aminopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC([NH3+])CC1

Specifications

Melting Point 44°C to 48°C
Quantity 1 g
UN Number UN3259
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material 4-Amino-1-Boc-piperidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.