missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-Boc-piperidine-4-carboxylic acid, 98%

Product Code. 11359755
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11359755

Brand: Thermo Scientific Alfa Aesar L19794.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 183673-71-4 | C11H20N2O4 | 244.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 183673-71-4
Molecular Formula C11H20N2O4
Molecular Weight (g/mol) 244.29
MDL Number MFCD01318728
InChI Key YNHLVALLAURVJF-UHFFFAOYSA-N
Synonym 1-boc-4-aminopiperidine-4-carboxylic acid, 4-amino-1-boc-piperidine-4-carboxylic acid, 4-amino-1-boc-isonipecotic acid, 4-amino-1-tert-butoxycarbonyl piperidine-4-carboxylic acid, h-pip boc-oh, 4,4-acp 1-n-boc, 4-amino-piperidine-1,4-dicarboxylic acid mono-tert-butyl ester, 1-boc-4-amino-4-piperidinecarboxylic acid, 4-amino-1-tert-butoxycarbonyl-4-piperidinecarboxylic acid, 4-amino-1-tert-butoxy carbonyl piperidine-4-carboxylic acid
PubChem CID 693797
SMILES CC(C)(C)OC(=O)N1CCC(N)(CC1)C(O)=O

Specifications

Melting Point ∽290°C (decomposition)
Quantity 1 g
Beilstein 7588844
IUPAC Name 4-amino-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-4-carboxylic acid
Formula Weight 244.29
Percent Purity 98%
Chemical Name or Material 4-Amino-1-Boc-piperidine-4-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.