missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-benzylpiperidine, 98%, Thermo Scientific Chemicals

Product Code. 10407570
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50541-93-0
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD00006504
InChI Key YUBDLZGUSSWQSS-UHFFFAOYSA-N
Synonym 4-amino-1-benzylpiperidine, 1-benzyl-4-aminopiperidine, 1-benzyl-piperidin-4-ylamine, n-benzyl-4-amino piperidine, 1-benzyl-4-piperidylamine, 4-piperidinamine, 1-phenylmethyl, 4-amino-1-n-benzylpiperidine, 4-amino-n-benzylpiperidine, 1-phenylmethyl-4-piperidinamine, n-benzyl-4-aminopiperidine
PubChem CID 415852
IUPAC Name 1-benzylpiperidin-4-amine
SMILES C1CN(CCC1N)CC2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 152°C (9.7 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5410 to 1.543
Quantity 5 g
Specific Gravity 0.93
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in ether
Formula Weight 190.29
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Amino-1-benzylpiperidine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Amino-1-benzylpiperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'