missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-benzylpiperidine, 98%

Product Code. 10113920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10113920 25 g Glass bottle 25g
10407570 5 g Glass bottle 5g
10052560 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10113920 Supplier Thermo Scientific Acros Supplier No. 187660250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50541-93-0 | C12H18N2 | 190.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50541-93-0
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD00006504
InChI Key YUBDLZGUSSWQSS-UHFFFAOYSA-N
Synonym 4-amino-1-benzylpiperidine, 1-benzyl-4-aminopiperidine, 1-benzyl-piperidin-4-ylamine, n-benzyl-4-amino piperidine, 1-benzyl-4-piperidylamine, 4-piperidinamine, 1-phenylmethyl, 4-amino-1-n-benzylpiperidine, 4-amino-n-benzylpiperidine, 1-phenylmethyl-4-piperidinamine, n-benzyl-4-aminopiperidine
PubChem CID 415852
IUPAC Name 1-benzylpiperidin-4-amine
SMILES C1CN(CCC1N)CC2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 152°C (9.7 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5410 to 1.543
Quantity 25 g
Specific Gravity 0.93
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in ether
Formula Weight 190.29
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Amino-1-benzylpiperidine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Amino-1-benzylpiperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.