missing translation for 'onlineSavingsMsg'
Learn More

4-Amidinopyridine hydrochloride, 98+%

Product Code. 11488893
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488893 5 g 5g
11478893 1 g 1g
11498893 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488893 Supplier Thermo Scientific Alfa Aesar Supplier No. A11050.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6345-27-3 | C6H8ClN3 | 157.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6345-27-3
Molecular Formula C6H8ClN3
Molecular Weight (g/mol) 157.60
MDL Number MFCD00040385
InChI Key IONKMFGAXKCLMI-UHFFFAOYSA-N
Synonym isonicotinimidamide hydrochloride, pyridine-4-carboximidamide hydrochloride, 4-amidinopyridine hydrochloride, isonicotinamidine hydrochloride, pyridine-4-carboxamidine hydrochloride, 4-pyridinecarboximidamide hydrochloride, 4-pyridinecarboximidamide, monohydrochloride, isonicotinamidine, hydrochloride, 4-amidino-pyridine hydrochloride, pyridine-4-carboxamidine, chloride
PubChem CID 2776844
IUPAC Name pyridine-4-carboximidamide;hydrochloride
SMILES Cl.NC(=N)C1=CC=NC=C1

Specifications

Melting Point ∼245°C (decomposition)
Quantity 5 g
Beilstein 3562678
Sensitivity Hygroscopic
Formula Weight 157.6
Percent Purity ≥98%
Chemical Name or Material 4-Amidinopyridine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.