missing translation for 'onlineSavingsMsg'
Learn More

4-Allyloxybenzaldehyde, 97%

Product Code. 11414834
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414834 50 g 50g
11404834 10 g 10g
11424834 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414834 Supplier Thermo Scientific Alfa Aesar Supplier No. A13773.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 40663-68-1 | C10H10O2 | 162.19 g/mol

4-Allyloxybenzaldehyde is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Allyloxybenzaldehyde is used as pharmaceutical intermediates.

Solubility
Immiscible in water.

Notes
Use with adequate ventilation. Store under inert atmosphere. Protect from moisture and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40663-68-1
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00014133
InChI Key TYNJQOJWNMZQFZ-UHFFFAOYSA-N
Synonym 4-allyloxybenzaldehyde, 4-allyloxy benzaldehyde, p-allyloxy benzaldehyde, p-allyloxybenzaldehyde, benzaldehyde, 4-2-propenyloxy, 4-prop-2-en-1-yloxy benzaldehyde, 4-prop-2-enyloxybenzaldehyde, 4-allyloxybenzaldehyd, acmc-20ampt
PubChem CID 95942
IUPAC Name 4-prop-2-enoxybenzaldehyde
SMILES C=CCOC1=CC=C(C=O)C=C1

Specifications

Density 1.058
Boiling Point 150°C to 152°C (18 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5695
Quantity 50 g
Beilstein 2206924
Sensitivity Air Sensitive
Solubility Information Immiscible in water.
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 4-Allyloxybenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.