missing translation for 'onlineSavingsMsg'
Learn More

4-Allyl-2,6-dimethoxyphenol, 98%

Product Code. 11439457
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439457 25 g 25g
11419457 1 g 1g
11429457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439457 Supplier Thermo Scientific Alfa Aesar Supplier No. B21981.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6627-88-9 | C11H14O3 | 194.23 g/mol

4-Allyl-2,6-dimethoxyphenol is used as a flavoring agent or adjuvant. Also used to produce 2,6-Dimethoxy-4-propyl-phenol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Allyl-2,6-dimethoxyphenol is used as a flavoring agent or adjuvant. Also used to produce 2,6-Dimethoxy-4-propyl-phenol.

Solubility
Soluble in fats. Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6627-88-9
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008655
InChI Key FWMPKHMKIJDEMJ-UHFFFAOYSA-N
Synonym 4-allyl-2,6-dimethoxyphenol, methoxyeugenol, 4-allylsyringol, phenol, 2,6-dimethoxy-4-2-propenyl, 2,6-dimethoxychavicol, phenol, 4-allyl-2,6-dimethoxy, unii-8vf00ywp89, 2,6-dimethoxy-4-allylphenol, 2,6-dimethoxy-4-prop-2-en-1-yl phenol, 4-hydroxy-3,5-dimethoxyallylbenzene
PubChem CID 226486
ChEBI CHEBI:86562
IUPAC Name 2,6-dimethoxy-4-prop-2-enylphenol
SMILES COC1=CC(CC=C)=CC(OC)=C1O

Specifications

Density 1.092
Boiling Point 168°C to 169°C (11 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.548
Quantity 25 g
Solubility Information Soluble in fats. Insoluble in water.
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material 4-Allyl-2,6-dimethoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.