missing translation for 'onlineSavingsMsg'
Learn More

4-Allyl-1,6-heptadien-4-ol, 99%

Product Code. 11485546
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485546 50 g 50g
11465546 2.5 g 2.5g
11475546 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485546 Supplier Thermo Scientific Alfa Aesar Supplier No. B20703.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10202-75-2 | C10H16O | 152.237 g/mol

A highly diastereoselective synthesis of homoallylic alcohols bearing up to two adjacent quaternary centers was achieved using these polysubstituted allylic reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A highly diastereoselective synthesis of homoallylic alcohols bearing up to two adjacent quaternary centers was achieved using these polysubstituted allylic reagents.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10202-75-2
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00039829
InChI Key SUXQWOWVXDXQSE-UHFFFAOYSA-N
Synonym 4-allyl-1,6-heptadien-4-ol, triallylcarbinol, 4-allylhepta-1,6-dien-4-ol, 4-allyl-1,6-heptadiene-4-ol, 1,6-heptadien-4-ol, 4-2-propenyl, 4-prop-2-en-1-yl hepta-1,6-dien-4-ol, triallyl carbinol, acmc-1byg9, 4-allyl-hepta-1,6-dien-4-ol, 1,6-heptadien-4-ol,4-2-propen-1-yl
PubChem CID 139107
IUPAC Name 4-prop-2-enylhepta-1,6-dien-4-ol
SMILES C=CCC(CC=C)(CC=C)O

Specifications

Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 152.24
Percent Purity 99%
Chemical Name or Material 4-Allyl-1,6-heptadien-4-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.