missing translation for 'onlineSavingsMsg'
Learn More

4-Acryloylmorpholine, 97%

Product Code. p-4258738 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
50mL
This item is not returnable. View return policy

Product Code. 11915491

Brand: Thermo Scientific Acros 439250500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5117-12-4 | C7H11NO2 | 141.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5117-12-4
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
InChI Key XLPJNCYCZORXHG-UHFFFAOYSA-N
SMILES C=CC(=O)N1CCOCC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -35.0°C
Color Colorless to Yellow
Density 1.1220g/mL
Boiling Point 158.0°C (50.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 50 mL
Specific Gravity 1.122
Synonym 4-acryloylmorpholine, 1-morpholinoprop-2-en-1-one, n-acrylylmorpholine, n-acryloylmorpholine, morpholine, 4-1-oxo-2-propenyl, poly acryloylmorpholine, acryloylmorpholine, acryloyl morpholine, 1-morpholin-4-yl prop-2-en-1-one, unii-k0y58p61ja
IUPAC Name 1-morpholin-4-ylprop-2-en-1-one
PubChem CID 98723
Formula Weight 141.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Acryloylmorpholine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Acryloylmorpholine, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P314-Get medical advice/attention if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.