missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylpyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-3735395
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1122-54-9
C7H7NO
121.14
MFCD00006433
WMQUKDQWMMOHSA-UHFFFAOYSA-N
4-acetylpyridine, methyl 4-pyridyl ketone, ethanone, 1-4-pyridinyl, ketone, methyl 4-pyridyl, 1-pyridin-4-yl ethan-1-one, 1-pyridin-4-yl ethanone, pyridine, 4-acetyl, 4-acetyl pyridine, 4-pyridyl methyl ketone, unii-g47437iow7
14282
1-pyridin-4-ylethanone
CC(=O)C1=CC=NC=C1
This item is not returnable. View return policy
1122-54-9
C7H7NO
121.14
MFCD00006433
WMQUKDQWMMOHSA-UHFFFAOYSA-N
4-acetylpyridine, methyl 4-pyridyl ketone, ethanone, 1-4-pyridinyl, ketone, methyl 4-pyridyl, 1-pyridin-4-yl ethan-1-one, 1-pyridin-4-yl ethanone, pyridine, 4-acetyl, 4-acetyl pyridine, 4-pyridyl methyl ketone, unii-g47437iow7
14282
1-pyridin-4-ylethanone
CC(=O)C1=CC=NC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 13.0°C to 16.0°C
Color Brown to Yellow
Density 1.0900g/mL
Boiling Point 212.0°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)COCH3
Refractive Index 1.5270 to 1.5310
Quantity 10 g
Beilstein 21, 279
Specific Gravity 1.09
Solubility Information Solubility in water: insoluble
Formula Weight 121.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Acetylpyridine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Acetylpyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'