missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylphenyl isothiocyanate, 98%, Thermo Scientific™

Product Code. 11342717
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342717 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11342717 Supplier Thermo Scientific Alfa Aesar Supplier No. L11927.03
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2131-57-9
Molecular Formula C9H7NOS
Molecular Weight (g/mol) 177.221
MDL Number MFCD00041100
InChI Key KAKXOESNHIUYJT-UHFFFAOYSA-N
Synonym 4-acetylphenyl isothiocyanate, 1-4-isothiocyanatophenyl ethanone, 4-acetylphenylisothiocyanate, 1-4-isothiocyanatophenyl ethan-1-one, acmc-20ampw, bromobenzyl-4-isothiocyanate, 4'-isothiocyanatoacetophenone, 4-acetylphenyl-isothiocyanate, phenylisothiocyanate, 4-acetyl, 1-4-isothiocyanato-phenyl-ethanone
PubChem CID 242779
IUPAC Name 1-(4-isothiocyanatophenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)N=C=S

Specifications

Melting Point 77°C to 81°C
Boiling Point 112°C (6 mmHg)
Odor Irritating
Quantity 1 g
UN Number UN2923
Beilstein 2207978
Sensitivity Moisture sensitive
Formula Weight 177.23
Percent Purity 98%
Chemical Name or Material 4-Acetylphenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.