missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyldiphenyl sulfide, 97%, Thermo Scientific™

Product Code. 11410571
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410571 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11410571 Supplier Thermo Scientific Alfa Aesar Supplier No. B20073.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10169-55-8
Molecular Formula C14H12OS
Molecular Weight (g/mol) 228.31
MDL Number MFCD00026227
InChI Key XUDYHODVSUXRPW-UHFFFAOYSA-N
Synonym 4-acetyldiphenyl sulfide, 1-4-phenylthio phenyl ethanone, 4-acetyl diphenyl sulfide, 4'-phenylthio acetophenone, 1-4-phenylsulfanyl phenyl ethanone, 1-4-phenylsulfanylphenyl ethanone, 1-4-phenylthio phenyl ethan-1-one, ethanone, 1-4-phenylthio phenyl, 1-acetyl-4-phenylthiobenzene
PubChem CID 66287
IUPAC Name 1-(4-phenylsulfanylphenyl)ethanone
SMILES CC(=O)C1=CC=C(SC2=CC=CC=C2)C=C1

Specifications

Melting Point 64°C to 68°C
Boiling Point 183°C to 184°C (3 mmHg)
Odor Odorless
Quantity 5 g
Beilstein 2049296
Formula Weight 228.32
Percent Purity 97%
Chemical Name or Material 4-Acetyldiphenyl sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.