missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbutyric Acid, TRC

Product Code. 30327544
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327544 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327544 Supplier Toronto Research Chemicals Supplier No. TRCA16844010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Acetylbutyric Acid
CAS 3128-06-1
Molecular Formula C6H10O3
Quantity 10 g
Synonym 4-Acetylbutyric Acid, 5-Ketocaproic Acid, 5-Ketohexanoic Acid, 5-Oxocaproic Acid, 5-Oxohexanoic Acid, NSC 5281, γ-Acetylbutyric Acid, δ-Ketocaproic Acid, δ-Ketohexanoic Acid, δ-Oxocaproic Acid
InChI Formula InChI=1S/C6H10O3/c1-5(7)3-2-4-6(8)9/h2-4H2,1H3,(H,8,9)
SMILES CC(=O)CCCC(=O)O
IUPAC Name 5-oxohexanoic acid
Molecular Weight (g/mol) 130.14
Formula Weight 130.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.