Learn More
4-Acetylbutyric acid, 97%
Description
4-Acetylbutyric acid is used in the synthesis of selective indomethacin analogues for AKR1C3 inhibition in the treatment of castrate-resistant prostate cancer. It is also used to prepare PARP inhibitors for the treatment of cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Acetylbutyric acid is used in the synthesis of selective indomethacin analogues for AKR1C3 inhibition in the treatment of castrate-resistant prostate cancer. It is also used to prepare PARP inhibitors for the treatment of cancer.
Solubility
Slightly soluble in water, chloroform and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 12°C to 14°C |
| Density | 1.1 |
| Boiling Point | 274°C to 275°C |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.4451 |
| Quantity | 25 g |
| Beilstein | 385840 |
| Solubility Information | Slightly soluble in water,chloroform and methanol. |
| SMILES | CC(=O)CCCC(=O)O |
| IUPAC Name | 5-oxohexanoic acid |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.