missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbutyric acid, 97%

Product Code. 11336736
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336736 5 g 5g
11346736 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336736 Supplier Thermo Scientific Alfa Aesar Supplier No. L03978.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3128-06-1 | C6H10O3 | 130.14 g/mol

4-Acetylbutyric acid is used in the synthesis of selective indomethacin analogues for AKR1C3 inhibition in the treatment of castrate-resistant prostate cancer. It is also used to prepare PARP inhibitors for the treatment of cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Acetylbutyric acid is used in the synthesis of selective indomethacin analogues for AKR1C3 inhibition in the treatment of castrate-resistant prostate cancer. It is also used to prepare PARP inhibitors for the treatment of cancer.

Solubility
Slightly soluble in water, chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3128-06-1
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00004412
InChI Key MGTZCLMLSSAXLD-UHFFFAOYSA-N
Synonym 4-acetylbutyric acid, 5-ketohexanoic acid, hexanoic acid, 5-oxo, 5-ketocaproic acid, 5-oxo-hexanoic acid, 4-acetylbutyricacid, 5-keto-n-caproic acid, 5-oxocaproic acid, delta-oxocaproic acid, delta-ketocaproic acid

Specifications

Melting Point 12°C to 14°C
Density 1.1
Boiling Point 274°C to 275°C
Flash Point >110°C (230°F)
Refractive Index 1.4451
Quantity 5 g
Beilstein 385840
Solubility Information Slightly soluble in water,chloroform and methanol.
SMILES CC(=O)CCCCC(=O)O
IUPAC Name 6-oxoheptanoic acid
PubChem CID 98810
Formula Weight 130.14
Percent Purity 97%
Chemical Name or Material 4-Acetylbutyric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.