missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetylbiphenyl, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 4-acetyl biphenyl |
| CAS | 92-91-1 |
| Molecular Formula | C14 H12 O |
| Quantity | 25 g |
| Synonym | (1-[1,1'-Biphenyl]-4-yl)ethanone, 1,1'-Biphenyl-4-yl methyl ketone, 1-(4-Phenylphenyl)ethanone, 1-Acetyl-4-phenylbenzene, 1-Biphenyl-4-ylethanone, 4-Acetyl-1,1'-biphenyl, 4-Acetylbiphenyl, 4-Biphenylyl methyl ketone, 4-Ethanoylbiphenyl, 4'-Phenylacetophenone, Methyl 4-biphenylyl ketone, NSC 1875, p-Acetylbiphenyl, p-Phenylacetophenone, Felbinac Imp. A (EP) |
| InChI Formula | InChI=1S/C14H12O/c1-11(15)12-7-9-14(10-8-12)13-5-3-2-4-6-13/h2-10H,1H3 |
| SMILES | CC(=O)c1ccc(cc1)c2ccccc2 |
| IUPAC Name | 1-(4-phenylphenyl)ethanone |
| Molecular Weight (g/mol) | 196.24 |
| Formula Weight | 196.0888 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction