missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbiphenyl, 98%

Product Code. 11497566
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11497566

Brand: Thermo Scientific Alfa Aesar A14622.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92-91-1 | C14H12O | 196.249 g/mol

4-Acetylbiphenyl is involved in the study of inactivation of 7-ethoxy-4-trifluoromethylcoumarin ortho-deethylase activity by various arylalkynes. Further, it is used in the preparation of Schiffs base.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Acetylbiphenyl is involved in the study of inactivation of 7-ethoxy-4-trifluoromethylcoumarin ortho-deethylase activity by various arylalkynes. Further, it is used in the preparation of Schiffs base.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-91-1
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.249
MDL Number MFCD00008749
InChI Key QCZZSANNLWPGEA-UHFFFAOYSA-N
Synonym 4-acetylbiphenyl, 4'-phenylacetophenone, 4-phenylacetophenone, 4-biphenylyl methyl ketone, p-phenylacetophenone, p-acetylbiphenyl, 1-1,1'-biphenyl-4-yl ethanone, acetophenone, 4'-phenyl, 1-4-phenylphenyl ethanone, ketone, 4-biphenylyl methyl
PubChem CID 7113
IUPAC Name 1-(4-phenylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 117°C to 123°C
Boiling Point 168°C (8 mmHg)
Quantity 500 g
Beilstein 1101615
Solubility Information Insoluble in water.
Formula Weight 196.25
Percent Purity 98%
Chemical Name or Material 4-Acetylbiphenyl

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.