Learn More
4-Acetylbenzoic acid, 98+%
Description
4-Acetylbenzoic acid was used in the preparation of an ester-derivative of paclitaxel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Acetylbenzoic acid was used in the preparation of an ester-derivative of paclitaxel.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 208°C to 212°C |
| Odor | Odorless |
| Quantity | 5 g |
| Beilstein | 2207355 |
| Solubility Information | Soluble in water. |
| Formula Weight | 164.16 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 4-Acetylbenzoic acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.