missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbenzoic acid, 98+%

Product Code. 11482278
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482278 1 g 1g
11492278 5 g 5g
11402288 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482278 Supplier Thermo Scientific Alfa Aesar Supplier No. A15239.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 586-89-0 | C9H8O3 | 164.16 g/mol

4-Acetylbenzoic acid was used in the preparation of an ester-derivative of paclitaxel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Acetylbenzoic acid was used in the preparation of an ester-derivative of paclitaxel.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-89-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00002561
InChI Key QBHDSQZASIBAAI-UHFFFAOYSA-N
Synonym benzoic acid, 4-acetyl, p-acetylbenzoic acid, 4-acetyl-benzoic acid, 4'-acetophenonecarboxylic acid, 4-acetylbezoic acid, 4-acetylbenzoci acid, 4'-carboxyacetohenone, pubchem3129, 4-acetyl benzoic acid
PubChem CID 11470
IUPAC Name 4-acetylbenzoic acid
SMILES CC(=O)C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 208°C to 212°C
Odor Odorless
Quantity 1 g
Beilstein 2207355
Solubility Information Soluble in water.
Formula Weight 164.16
Percent Purity ≥98%
Chemical Name or Material 4-Acetylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.