missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbenzenesulfonamide, 97%

Product Code. 15415677
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415677 5 g 5g
15405677 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415677 Supplier Thermo Scientific Alfa Aesar Supplier No. H32901.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1565-17-9 | C8H9NO3S | 199.22 g/mol

4-acetylbenzenesulfonamide and acetohexamide which is used to characterize the pharmacokinetics of a reversible drug—metabolite system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-acetylbenzenesulfonamide and acetohexamide which is used to characterize the pharmacokinetics of a reversible drug—metabolite system.

Solubility
soluble in water( 2650 mg/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1565-17-9
Molecular Formula C8H9NO3S
Molecular Weight (g/mol) 199.22
MDL Number MFCD00792524
InChI Key CSATVXJBGFVJES-UHFFFAOYSA-N
Synonym 4-acetylbenzenesulphonamide, p-acetylbenzenesulfonamide, 4-acetyl-benzenesulfonamide, 4-acetylbenzene-1-sulfonamide, benzenesulfonamide, 4-acetyl, p-acetobenzenesulfonamide, benzenesulfonamide, p-acetyl-6ci,7ci,8ci, 4-acetyl-benzenesulfonamid, p-acetylbenzene sulfonamide, #
PubChem CID 74065
IUPAC Name 4-acetylbenzenesulfonamide
SMILES CC(=O)C1=CC=C(C=C1)S(N)(=O)=O

Specifications

Melting Point 177°C to 179°C
Quantity 5 g
Solubility Information soluble in water( 2650mg/L).
Formula Weight 199.23
Percent Purity 97%
Chemical Name or Material 4-Acetylbenzenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.