missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetylbenzenesulfonamide, 97%
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
4-acetylbenzenesulfonamide and acetohexamide which is used to characterize the pharmacokinetics of a reversible drug—metabolite system.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-acetylbenzenesulfonamide and acetohexamide which is used to characterize the pharmacokinetics of a reversible drug—metabolite system.
Solubility
soluble in water( 2650 mg/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
4-acetylbenzenesulfonamide and acetohexamide which is used to characterize the pharmacokinetics of a reversible drug—metabolite system.
Solubility
soluble in water( 2650 mg/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 177°C to 179°C |
| Quantity | 1 g |
| Solubility Information | soluble in water( 2650mg/L). |
| Formula Weight | 199.23 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Acetylbenzenesulfonamide |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction