missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbenzeneboronic acid, 98%

Product Code. 11470847
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470847 25 g 25g
11450847 1 g 1g
11460847 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470847 Supplier Thermo Scientific Alfa Aesar Supplier No. B23234.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 149104-90-5 | C8H9BO3 | 163.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149104-90-5
Molecular Formula C8H9BO3
Molecular Weight (g/mol) 163.97
MDL Number MFCD01074667
InChI Key OBQRODBYVNIZJU-UHFFFAOYSA-N
Synonym 4-acetylphenyl boronic acid, p-acetylphenylboronic acid, 4-acetylbenzeneboronic acid, 1-4-dihydroxyboranyl phenyl ethan-1-one, boronic acid, 4-acetylphenyl, 4acetylphenylboronic acid, 4-actylphenylboronic acid, 4acetylbenzeneboronic acid, p-acetylbenzeneboronic acid
PubChem CID 3702122
IUPAC Name (4-acetylphenyl)boronic acid
SMILES CC(=O)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point ∼240°C (decomposition)
Quantity 25 g
Beilstein 7869162
Formula Weight 163.97
Percent Purity 98%
Chemical Name or Material 4-Acetylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.