missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylamino-3-hydroxybenzoic Acid Ethyl Ester, TRC

Product Code. 17982285
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982285 10 mg 10mg
17953705 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17982285 Supplier Toronto Research Chemicals Supplier No. TRCA16825010MG
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1346604-18-9
Molecular Formula C11N13NO4
Molecular Weight (g/mol) 223.23
Synonym Benzoic acid, 4-(acetylamino)-3-hydroxy-, ethyl ester, Ethyl 4-acetamido-3-hydroxybenzoate, 4-Acetylamino-3-hydroxybenzoic acid ethyl ester
IUPAC Name ethyl 4-acetamido-3-hydroxybenzoate
SMILES CCOC(=O)c1ccc(NC(=O)C)c(O)c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H13NO4/c1-3-16-11(15)8-4-5-9(10(14)6-8)12-7(2)13/h4-6,14H,3H2,1-2H3,(H,12,13)
Formula Weight 223.0845
Percent Purity >95%
Chemical Name or Material 4-Acetylamino-3-hydroxybenzoic Acid Ethyl Ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.