missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyl-2-fluorobiphenyl, TRC

Product Code. 18092930
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18092930 500 mg 500mg
18033020 50 mg 50mg
18020320 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18092930 Supplier Toronto Research Chemicals Supplier No. TRCA168850500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42771-79-9
Molecular Formula C14 H11 F O
Molecular Weight (g/mol) 214.23
Synonym Ethanone, 1-(2-fluoro[1,1'-biphenyl]-4-yl)- (9CI), 3'-Fluoro-4'-phenylacetophenone, 4-Acetyl-2-fluorobiphenyl, 1-(2-Fluorobiphenyl-4-yl)ethanone, Flurbiprofen Impurity D
IUPAC Name 1-(3-fluoro-4-phenylphenyl)ethanone
SMILES CC(=O)c1ccc(c(F)c1)c2ccccc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H11FO/c1-10(16)12-7-8-13(14(15)9-12)11-5-3-2-4-6-11/h2-9H,1H3
Formula Weight 214.0794
Percent Purity >95%
Chemical Name or Material 4-Acetyl 2-Fluorobiphenyl
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.