missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxystyrene, 95%, stab.

Product Code. 11332517
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332517 2 g 2g
11342517 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332517 Supplier Thermo Scientific Alfa Aesar Supplier No. L11601.04
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2628-16-2 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2628-16-2
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00075734
InChI Key JAMNSIXSLVPNLC-UHFFFAOYSA-N
Synonym 4-acetoxystyrene, 4-vinylphenyl acetate, 4-ethenylphenol acetate, p-acetoxystyrene, phenol, 4-ethenyl-, acetate, p-vinylphenol acetate, unii-7s21904ayn, phenol, 4-ethenyl-, 1-acetate, 4-ethenylphenyl acetate, acetic acid 4-vinylphenyl ester
PubChem CID 75821
IUPAC Name (4-ethenylphenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)C=C

Specifications

Melting Point 7°C to 8°C
Density 1.06
Boiling Point 259°C to 260°C
Flash Point 87°C (188°F)
Refractive Index 1.538
Quantity 2 g
Beilstein 1862793
Formula Weight 162.19
Percent Purity 95%
Chemical Name or Material 4-Acetoxystyrene, stabilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.