missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxybenzoic acid, 98+%

Product Code. 11339439
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
500 g
Unit Size:
250g
25g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339439 250 g 250g
11450143 25 g 25g
11329439 50 g 50g
11460143 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11339439 Supplier Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A11350.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 2345-34-8 | C9H8O4 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 2345-34-8
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00002540
InChI Key GDBUZIKSJGRBJP-UHFFFAOYSA-N
Synonym 4-acetoxybenzoic acid, p-acetoxybenzoic acid, 4-acetyloxy benzoic acid, 4-carboxyphenyl acetate, benzoic acid, 4-acetyloxy, p-hydroxybenzoic acid acetate, p-acetyloxybenzoic acid, p-carboxyphenyl acetate, benzoic acid, p-hydroxy-, acetate, 4-acetoxy benzoic acid
PubChem CID 16865
ChEBI CHEBI:86560
IUPAC Name 4-acetyloxybenzoic acid
SMILES CC(=O)OC1=CC=C(C=C1)C(O)=O

Specifikationer

Melting Point 192°C to 195°C
Quantity 250 g
Beilstein 1871997
Formula Weight 180.15
Percent Purity ≥98%
Chemical Name or Material 4-Acetoxybenzoic acid

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.