missing translation for 'onlineSavingsMsg'
Learn More

4'-Acetoxyacetophenone, 99%

Product Code. 11435956
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435956 100 g 100g
11425956 25 g 25g
11445956 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435956 Supplier Thermo Scientific Alfa Aesar Supplier No. B22284.22
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13031-43-1 | C10H10O3 | 178.19 g/mol

4'-Acetoxyacetophenone it can react with ethane-1,2-diol to get 2-(4-acetoxyphenyl)-2-methyl-1,3-dioxolane with yield is 67 %.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Acetoxyacetophenone it can react with ethane-1,2-diol to get 2-(4-acetoxyphenyl)-2-methyl-1,3-dioxolane with yield is 67 %.

Solubility
Slightly soluble in water(4450 mg/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13031-43-1
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00017229
InChI Key SMIOEQSLJNNKQF-UHFFFAOYSA-N
Synonym 4'-acetoxyacetophenone, p-acetoxyacetophenone, 4-acetoxyacetophenone, p-acetylphenyl acetate, 4-acetylphenyl acetate, unii-eo1xk390jf, ethanone, 1-4-acetyloxy phenyl, 4-acetoxy acetophenone, 4-acetoxy-acetophenone, eo1xk390jf
PubChem CID 83063
ChEBI CHEBI:86558
IUPAC Name (4-acetylphenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 51°C to 54°C
Boiling Point 165°C to 170°C (14 mmHg)
Quantity 100 g
Beilstein 2093080
Solubility Information Slightly soluble in water(4450mg/L).
Formula Weight 178.19
Percent Purity 99%
Chemical Name or Material 4'-Acetoxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.