missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxy-3-methoxybenzaldehyde, 98%

Product Code. 10288473 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10288473 25 g Glass bottle 25g
10530184 5 g Glass bottle 5g
10350864 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10288473 Supplier Thermo Scientific Acros Supplier No. 385940250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 881-68-5 | C10H10O4 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 881-68-5
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00003362
InChI Key PZSJOBKRSVRODF-UHFFFAOYSA-N
Synonym vanillin acetate, 4-acetoxy-3-methoxybenzaldehyde, acetovanillin, acetylvanillin, vanillin, acetate, 4-o-acetylvanillin, o-acetylvanillin, benzaldehyde, 4-acetyloxy-3-methoxy, 3-methoxy-4-acetoxybenzaldehyde, acetyl vanillin
PubChem CID 61229
ChEBI CHEBI:86956
IUPAC Name (4-formyl-2-methoxyphenyl) acetate
SMILES CC(=O)OC1=C(C=C(C=C1)C=O)OC

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 73°C to 78°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(CHO)OC(=O)CH3
Quantity 25 g
Formula Weight 194.19
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetoxy-3-methoxybenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.